Merck
  • Dynamic epigenetic regulation of glioblastoma tumorigenicity through LSD1 modulation of MYC expression.
Dynamic epigenetic regulation of glioblastoma tumorigenicity through LSD1 modulation of MYC expression.
Proceedings of the National Academy of Sciences of the United States of America (2015-07-15)
David Kozono, Jie Li, Masayuki Nitta, Oltea Sampetrean, David Gonda, Deepa S Kushwaha, Dmitry Merzon, Valya Ramakrishnan, Shan Zhu, Kaya Zhu, Hiroko Matsui, Olivier Harismendy, Wei Hua, Ying Mao, Chang-Hyuk Kwon, Hideyuki Saya, Ichiro Nakano, Donald P Pizzo, Scott R VandenBerg, Clark C Chen
ABSTRACT
The available evidence suggests that the lethality of glioblastoma is driven by small subpopulations of cells that self-renew and exhibit tumorigenicity. It remains unclear whether tumorigenicity exists as a static property of a few cells or as a dynamically acquired property. We used tumor-sphere and xenograft formation as assays for tumorigenicity and examined subclones isolated from established and primary glioblastoma lines. Our results indicate that glioblastoma tumorigenicity is largely deterministic, yet the property can be acquired spontaneously at low frequencies. Further, these dynamic transitions are governed by epigenetic reprogramming through the lysine-specific demethylase 1 (LSD1). LSD depletion increases trimethylation of histone 3 lysine 4 at the avian myelocytomatosis viral oncogene homolog (MYC) locus, which elevates MYC expression. MYC, in turn, regulates oligodendrocyte lineage transcription factor 2 (OLIG2), SRY (sex determining region Y)-box 2 (SOX2), and POU class 3 homeobox 2 (POU3F2), a core set of transcription factors required for reprogramming glioblastoma cells into stem-like states. Our model suggests epigenetic regulation of key transcription factors governs transitions between tumorigenic states and provides a framework for glioblastoma therapeutic development.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Nol3
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Myc
Sigma-Aldrich
MISSION® esiRNA, targeting human MYC
Sigma-Aldrich
MISSION® esiRNA, targeting human NOL3