Merck
  • Mechanical properties and in vivo healing evaluation of a novel Centella asiatica-loaded hydrocolloid wound dressing.
Mechanical properties and in vivo healing evaluation of a novel Centella asiatica-loaded hydrocolloid wound dressing.
International journal of pharmaceutics (2015-05-31)
Sung Giu Jin, Kyung Soo Kim, Abid Mehmood Yousaf, Dong Wuk Kim, Sun Woo Jang, Mi-Won Son, Young Hun Kim, Chul Soon Yong, Jong Oh Kim, Han-Gon Choi
ABSTRACT
To develop a novel sodium alginate based Centella asiatica (CA)-loaded hydrocolloid wound dressing (HCD) providing excellent mechanical properties and improved wound healing, numerous CA-loaded HCDs were prepared with various ingredients using the hot melting method. The effect of sodium alginate, styrene-isoprene-styrene copolymer (SIS) and petroleum hydrocarbon resin (PHR) on the mechanical properties of CA-loaded HCDs was investigated. The effect of disintegrants on swelling and drug release was assessed. Moreover, the in vivo wound healing potentials of the selected CA-loaded HCD in various wound models such as abrasion, excision and infection were evaluated in comparison with the commercial product. Polyisobutylene and SIS hardly affected the mechanical properties, but PHR improved the tensile strength and elongation at break. Disintegrants such as croscarmellose sodium, sodium starch glycolate and crospovidone improved the swelling ratio of the CA-loaded HCD. Furthermore, the CA-loaded HCD without croscarmellose sodium poorly released the drug, but that with 2% croscarmellose sodium showed about 27% drug release in 24h. In particular, the CA-loaded HCD composed of CA/polyisobutylene/SIS/PHR/liquid paraffin/sodium alginate/croscarmellose sodium at the weight ratio of 1/8/25/25/12/27/2 furnished excellent mechanical properties and drug release. As compared with the commercial product, it offered improved healing effects in excision, infection and abrasion type wounds in rats. Thus, this novel CA-loaded HCD could be a potential candidate for the treatment of various wounds.
Sigma-Aldrich
Streptozocin, ≥75% α-anomer basis, ≥98% (HPLC), powder
Sigma-Aldrich
Streptozocin, Vetec, reagent grade, 98%, powder
Sigma-Aldrich
Styrene, ReagentPlus®, contains 4-tert-butylcatechol as stabilizer, ≥99%