Merck
  • Multilocus genotypic characterization of Escherichia coli O157:H7 recovered from food sources.
Multilocus genotypic characterization of Escherichia coli O157:H7 recovered from food sources.
Epidemiology and infection (2014-11-28)
M M Elhadidy, W F Elkhatib
ABSTRACT
Escherichia coli O157:H7 strains (n = 33) recovered from different food sources in Egypt were characterized using molecular assays to identify strain genotypes associated with various levels of pathogenic potential. Genotypic characterization included: lineage-specific polymorphism assay (LSPA-6), Shiga-toxin-encoding bacteriophage insertion site assay (SBI), clade 8 typing, Tir (A255 T) polymorphism, and variant analysis of Shiga toxin 2 gene (Stx 2a and Stx 2c), and anti-terminator Q genes (Q 933 and Q 21). Genotypes LI/II (76%), SBI 1 (60·6%), clade 8 (69·7%), Tir (255 T) (72·7%) and Stx 2c (45·5%) were found to be significantly more frequent compared to other genetic markers in the strains analysed. Multivariable analysis revealed a significant association between LPSA-6 and clade types as well as Tir (A255 T). To the best of our knowledge, this is the first study to report the characterization of these genetic markers in E. coli O157:H7 strains in the Middle East and Africa.
Sigma-Aldrich
Trichostatin A , Vetec, reagent grade, from Streptomyces sp., ≥98%