Merck
  • Sigma-1 receptor mediates acquisition of alcohol drinking and seeking behavior in alcohol-preferring rats.
Sigma-1 receptor mediates acquisition of alcohol drinking and seeking behavior in alcohol-preferring rats.
Behavioural brain research (2015-04-08)
Angelo Blasio, Marta Valenza, Malliga R Iyer, Kenner C Rice, Luca Steardo, T Hayashi, Pietro Cottone, Valentina Sabino
ABSTRACT
Sigma-1 receptor (Sig-1R) has been proposed as a novel therapeutic target for drug and alcohol addiction. We have shown previously that Sig-1R agonists facilitate the reinforcing effects of ethanol and induce binge-like drinking, while Sig-1R antagonists on the other hand block excessive drinking in genetic and environmental models of alcoholism, without affecting intake in outbred non-dependent rats. Even though significant progress has been made in understanding the function of Sig-1R in alcohol reinforcement, its role in the early and late stage of alcohol addiction remains unclear. Administration of the selective Sig-1R antagonist BD-1063 dramatically reduced the acquisition of alcohol drinking behavior as well as the preference for alcohol in genetically selected TSRI Sardinian alcohol preferring (Scr:sP) rats; the treatment had instead no effect on total fluid intake, food intake or body weight gain, proving selectivity of action. Furthermore, BD-1063 dose-dependently decreased alcohol-seeking behavior in rats trained under a second-order schedule of reinforcement, in which responding is maintained by contingent presentation of a conditioned reinforcer. Finally, an innate elevation in Sig-1R protein levels was found in the nucleus accumbens of alcohol-preferring Scr:sP rats, compared to outbred Wistar rats, alteration which was normalized by chronic, voluntary alcohol drinking. Taken together these findings demonstrate that Sig-1R blockade reduces the propensity to both acquire alcohol drinking and to seek alcohol, and point to the nucleus accumbens as a potential key region for the effects observed. Our data suggest that Sig-1R antagonists may have therapeutic potential in multiple stages of alcohol addiction.
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, Vetec, reagent grade, 98%