Merck
  • ZnO/Ag/CdO nanocomposite for visible light-induced photocatalytic degradation of industrial textile effluents.
ZnO/Ag/CdO nanocomposite for visible light-induced photocatalytic degradation of industrial textile effluents.
Journal of colloid and interface science (2015-05-04)
R Saravanan, M Mansoob Khan, Vinod Kumar Gupta, E Mosquera, F Gracia, V Narayanan, A Stephen
ABSTRACT
A ternary ZnO/Ag/CdO nanocomposite was synthesized using thermal decomposition method. The resulting nanocomposite was characterized by X-ray diffraction, field emission scanning electron microscopy, transmission electron microscopy, UV-Vis spectroscopy, and X-ray photoelectron spectroscopy. The ZnO/Ag/CdO nanocomposite exhibited enhanced photocatalytic activity under visible light irradiation for the degradation of methyl orange and methylene blue compared with binary ZnO/Ag and ZnO/CdO nanocomposites. The ZnO/Ag/CdO nanocomposite was also used for the degradation of the industrial textile effluent (real sample analysis) and degraded more than 90% in 210 min under visible light irradiation. The small size, high surface area and synergistic effect in the ZnO/Ag/CdO nanocomposite is responsible for high photocatalytic activity. These results also showed that the Ag nanoparticles induced visible light activity and facilitated efficient charge separation in the ZnO/Ag/CdO nanocomposite, thereby improving the photocatalytic performance.
Sigma-Aldrich
Cadmium(II) acetate, LR, ≥98%
Sigma-Aldrich
Cadmium(II) acetate, anhydrous, 99.995%
Cadmium(II) acetate, SAFC Hitech®, anhydrous, 99.995%