Merck
  • No effect of oral L-tryptophan or alpha-lactalbumin on total tryptophan levels in breast milk.
No effect of oral L-tryptophan or alpha-lactalbumin on total tryptophan levels in breast milk.
European neuropsychopharmacology : the journal of the European College of Neuropsychopharmacology (2015-04-01)
Yekta Dowlati, Arun V Ravindran, Maxim Maheux, Meir Steiner, Donna E Stewart, Jeffrey H Meyer
ABSTRACT
Postpartum depression (PPD) is the most common complication of childbearing with a 13% prevalence rate. Sleep disturbances are also common, particularly during early postpartum. In theory, l-tryptophan could improve sleep and reduce depressed mood in early postpartum; however, the first step in clinical development of tryptophan for use in postpartum is to measure the effect of oral l-tryptophan on its concentrations in breast milk, which is presently unknown. The aims were to investigate the effect of oral l-tryptophan and alpha-lactalbumin, a protein with high tryptophan concentration, on total and free tryptophan levels in breast milk and plasma, and to compare free tryptophan levels in breast milk with those in common infant formulas. Thirty healthy breastfeeding women were randomly allocated to receive 2g or 4g of l-tryptophan, or, 20g or 40g of alpha-lactalbumin or no supplement. Free tryptophan levels were also measured in 12 different infant formulas. Total tryptophan in breast milk was unaffected by oral administration of l-tryptophan or alpha-lactalbumin (repeated measures of ANOVA (rANOVA), group effect: p=0.93). Both l-tryptophan and alpha-lactalbumin were associated with greater free tryptophan levels in breast milk (rANOVA, group effect: p<0.001) (representing 2% of total tryptophan), but these concentrations were within the range of commonly used infant formulas. In contrast to most sleep inducing medications, l-tryptophan does not affect its total concentration in breast milk. These results support further investigation of dietary l-tryptophan and alpha-lactalbumin as part of a dietary supplementation approach to address sleep disturbances in postpartum and reduce risk of PPD.
Sigma-Aldrich
DL-Tryptophan, ≥99% (HPLC)
Sigma-Aldrich
DL-Tryptophan, ≥99% (HPLC)