Merck
  • Impaired extracellular matrix structure resulting from malnutrition in ovariectomized mature rats.
Impaired extracellular matrix structure resulting from malnutrition in ovariectomized mature rats.
Histochemistry and cell biology (2015-07-27)
Thaqif El Khassawna, Wolfgang Böcker, Katharina Brodsky, David Weisweiler, Parameswari Govindarajan, Marian Kampschulte, Ulrich Thormann, Anja Henss, Marcus Rohnke, Natali Bauer, Robert Müller, Andreas Deutsch, Anita Ignatius, Lutz Dürselen, Alexander Langheinrich, Katrin S Lips, Reinhard Schnettler, Christian Heiss
ABSTRACT
Bone loss is a symptom related to disease and age, which reflects on bone cells and ECM. Discrepant regulation affects cell proliferation and ECM localization. Rat model of osteoporosis (OVX) was investigated against control rats (Sham) at young and old ages. Biophysical, histological and molecular techniques were implemented to examine the underlying cellular and extracellular matrix changes and to assess the mechanisms contributing to bone loss in the context of aging and the widely used osteoporotic models in rats. Bone loss exhibited a compromised function of bone cells and infiltration of adipocytes into bone marrow. However, the expression of genes regulating collagen catabolic process and adipogenesis was chronologically shifted in diseased bone in comparison with aged bone. The data showed the involvement of Wnt signaling inhibition in adipogenesis and bone loss due to over-expression of SOST in both diseased and aged bone. Further, in the OVX animals, an integrin-mediated ERK activation indicated the role of MAPK in osteoblastogenesis and adipogenesis. The increased PTH levels due to calcium and estrogen deficiency activated osteoblastogenesis. Thusly, RANKL-mediated osteoclastogenesis was initiated. Interestingly, the data show the role of MEPE regulating osteoclast-mediated resorption at late stages in osteoporotic bone. The interplay between ECM and bone cells change tissue microstructure and properties. The involvement of Wnt and MAPK pathways in activating cell proliferation has intriguing similarities to oncogenesis and myeloma. The study indicates the importance of targeting both pathways simultaneously to remedy metabolic bone diseases and age-related bone loss.
Sigma-Aldrich
Hydrogen peroxide solution, AR, contains sodium pyrophosphate as stabilizer, ≥30 % (w/v)
Sigma-Aldrich
Sodium acetate, suitable for HPLC, ≥99.5%
Sigma-Aldrich
N,N-Dimethylformamide, LR, ≥99%
Sigma-Aldrich
Sodium acetate, LR, anhydrous, ≥98%
Sigma-Aldrich
N,N-Dimethylformamide, AR, ≥99.5%
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.5%
Sigma-Aldrich
Hydrogen peroxide solution, LR, contains sodium pyrophosphate as stabilizer, ≥30 % (w/v)
Sigma-Aldrich
Hydrogen peroxide solution, LR, contains sodium pyrophosphate as stabilizer, ≥50 % (w/v)
Sigma-Aldrich
Sodium acetate, Vetec, reagent grade, 98%
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Hydrogen peroxide solution, semiconductor grade MOS PURANAL (Honeywell 17937), ≥30%
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
Aluminum sulfate, 99.99% trace metals basis
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Hydrogen peroxide solution, semiconductor grade VLSI PURANAL (Honeywell 17606), ≥30%
Sigma-Aldrich
Xylazine, ≥99%
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
N,N-Dimethylformamide, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%