Merck
  • Professional phagocytic granulocyte-derived PGD2 regulates the resolution of inflammation in fish.
Professional phagocytic granulocyte-derived PGD2 regulates the resolution of inflammation in fish.
Developmental and comparative immunology (2015-06-02)
Victoria Gómez-Abellán, Jana Montero, Azucena López-Muñoz, Antonio Figueras, Marta Arizcun, Victoriano Mulero, María P Sepulcre
ABSTRACT
Prostaglandins (PGs) play a key role in the development on the immune response through the regulation of both pro- and anti-inflammatory processes. PGD(2) can be either pro- or anti-inflammatory depending on the inflammatory milieu. Prostaglandin D synthase (PGDS) is the enzyme responsible for the conversion of PGH(2) to PGD(2). In mammals, two types of PGDS synthase have been described, the hematopoietic (H-PGDS) and the lipocalin (L-PGDS). In the present study we describe the existence of two orthologs of the mammalian L-PGDS (PGDS1 and PGDS2) in the gilthead seabream and characterize their gene expression profiles and biological activity. The results showed a dramatic induction of the gene coding for PGDS1 in acidophilic granulocytes (AGs), which are functionally equivalent to mammalian neutrophils, after a prolonged in vitro activation with different pathogen associated molecular patterns (PAMPs). In contrast PGDS2 was not expressed in these cells. The functional relevance of the induction of PGDS1 in AGs was confirmed by the ability of these cells to release PGD(2) upon PAMP stimulation. To gain further insight into the role of PGD(2) in the resolution of inflammation in fish, we examined the ability of PGD(2) or its cyclopentenone derivates (cyPGs) to modulate the main functional activities of AGs. It was found that both PGD(2) and cyPGs inhibited the production of reactive oxygen species and downregulated the transcript levels of the gene encoding interleukin-1β. Taken together, these results demonstrate that the use of PGD(2) and its metabolites in the resolution of inflammation was established before the divergence of fish from tetrapods more than 450 million years ago and support a critical role for granulocytes in the resolution of inflammation in vertebrates.
Sigma-Aldrich
Prostaglandin D2, ≥95%, synthetic
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Luminol, 97%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Luminol, ≥97% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02 N
Sigma-Aldrich
Ethylenediaminetetraacetic acid, Vetec, reagent grade, 98%