Merck
  • Dioscin ameliorates cerebral ischemia/reperfusion injury through the downregulation of TLR4 signaling via HMGB-1 inhibition.
Dioscin ameliorates cerebral ischemia/reperfusion injury through the downregulation of TLR4 signaling via HMGB-1 inhibition.
Free radical biology & medicine (2015-03-17)
Xufeng Tao, Xiance Sun, Lianhong Yin, Xu Han, Lina Xu, Yan Qi, Youwei Xu, Hua Li, Yuan Lin, Kexin Liu, Jinyong Peng
ABSTRACT
We previously reported the promising effect of dioscin against hepatic ischemia/reperfusion (I/R) injury, but its effect on cerebral I/R injury remains unknown. In this work, an in vitro oxygen-glucose deprivation and reoxygenation (OGD/R) model and an in vivo middle cerebral artery occlusion (MCAO) model were used. The results indicated that dioscin clearly protected PC12 cells and primary cortical neurons against OGD/R insult and significantly prevented cerebral I/R injury. Further research demonstrated that dioscin-induced neuroprotection was accompanied by a significant inhibition in the expression and the nuclear to cytosolic translocation of HMGB-1, reflected by decreased TLR4 expression. Blockade of the TLR4/MyD88/TRAF6 signaling pathway by dioscin inhibited NF-κB and AP-1 transcriptional activities, MAPK and STAT3 phosphorylation, and pro-inflammatory cytokine responses, and upregulated the levels of anti-inflammatory factors. In addition, small interfering RNA (siRNA) and overexpressed genes of HMGB-1 and TLR4 were applied in in vitro experiments, respectively, and the results further confirmed that dioscin showed an efficient neuroprotection because of its inhibiting effects on HMGB-1/TLR4 signaling and subsequent suppressing inflammation. These findings provide new insights that will aid in elucidating the effect of dioscin against cerebral I/R injury and support the development of dioscin as a potential treatment for ischemic stroke.
Sigma-Aldrich
2,3,5-Triphenyltetrazolium chloride, ≥98.0% (HPLC)