Merck
  • Biological Effects of the Herbal Plant-Derived Phytoestrogen Bavachin in Primary Rat Chondrocytes.
Biological Effects of the Herbal Plant-Derived Phytoestrogen Bavachin in Primary Rat Chondrocytes.
Biological & pharmaceutical bulletin (2015-08-04)
Gyeong-Je Lee, In-A Cho, Kyeong-Rok Kang, Do Kyung Kim, Hong-Moon Sohn, Jae-Won You, Ji-Su Oh, Yo-Seob Seo, Sang-Joun Yu, Jae-Seek You, Chun Sung Kim, Su-Gwan Kim, Hee-Jeong Im, Jae-Sung Kim
ABSTRACT
The aim of this study was to examine the anabolic and anticatabolic functions of bavachin in primary rat chondrocytes. With bavachin treatment, chondrocytes survived for 21 d without cell proliferation, and the proteoglycan content and extracellular matrix increased. Short-term monolayer culture of chondrocytes showed that gene induction of both aggrecan and collagen type II, major extracellular matrix components, was significantly upregulated by bavachin. The expression and activities of cartilage-degrading enzymes such as matrix metalloproteinases and a disintegrin and metalloproteinase with thrombospondin motifs were inhibited significantly by bavachin, while tissue inhibitors of metalloprotease were significantly upregulated. Bavachin inhibits the expression of inducible nitric oxide synthase, a representative catabolic factor, and downregulated the expression of nitric oxide, cyclooxygenase-2, and prostaglandin E2 in a dose-dependent manner in chondrocytes. Our results suggest that the bavachin has anabolic and potent anticatabolic biological effects on chondrocytes, which may have considerable promise in treating articular cartilage degeneration in the future.
Sigma-Aldrich
4-Chromanone, 97%