Merck
  • Translocation t(10;14)(q11.2:q22.1) fusing the kinetin to the RET gene creates a novel rearranged form (PTC8) of the RET proto-oncogene in radiation-induced childhood papillary thyroid carcinoma.
Translocation t(10;14)(q11.2:q22.1) fusing the kinetin to the RET gene creates a novel rearranged form (PTC8) of the RET proto-oncogene in radiation-induced childhood papillary thyroid carcinoma.
Cancer research (2000-06-13)
K Salassidis, J Bruch, H Zitzelsberger, E Lengfelder, A M Kellerer, M Bauchinger
ABSTRACT
Evaluation of 20 cases of radiation-induced childhood papillary thyroid carcinoma using fluorescence in situ hybridization demonstrated the presence of clonal translocations affecting the RET locus. Semiquantitative reverse transcription-PCR indicated overexpression of the RET tyrosine kinase (TK) domain in four cases. In two cases, the RET rearrangements PTC6 and PTC7 were identified and assigned to balanced translocations t(7;10)(q32;q11.2) and t(1;10)(p13;q11.2), respectively. In one case with a balanced translocation t(10;14)(q11.2;q22.1), 5' rapid amplification of cDNA ends revealed a novel type of RET oncogenic activation (PTC8), arising from a fusion of the 5' part of the kinectin (KTN1) gene to the TK domain of the RET gene. The presence of coiled-coil domains in the resulting ktn1/ret fusion protein suggests ligand-independent dimerization and thus constitutive activation of the ret TK domain.