Merck
  • GPR18 in microglia: implications for the CNS and endocannabinoid system signalling.
GPR18 in microglia: implications for the CNS and endocannabinoid system signalling.
British journal of pharmacology (2012-05-09)
D McHugh
ABSTRACT
A review of what is presently known about the G protein coupled receptor GPR18 in terms of its expression and distribution, pharmacology and potential implications for central nervous system and endocannabinoid system signalling. LINKED ARTICLES This article is part of a themed section on Cannabinoids. To view the other articles in this section visit http://dx.doi.org/10.1111/bph.2012.167.issue-8.