Merck
  • Inhibition of ZNF746 suppresses invasion and epithelial to mesenchymal transition in H460 non-small cell lung cancer cells.
Inhibition of ZNF746 suppresses invasion and epithelial to mesenchymal transition in H460 non-small cell lung cancer cells.
Oncology reports (2013-10-23)
Bonglee Kim, Eun Jung Sohn, Ji Hoon Jung, Eun Ah Shin, Ok Heui You, Jinwon Im, Sung-Hoon Kim
ABSTRACT
Although ZNF746, also known as Parkin-interacting substrate (PARIS), has been reported to suppress peroxisome proliferator-activated receptor γ coactivator-1α (PGC-1α) and its target gene NRF-1 leading to the neurodegeneration in Parkinson's disease, its function in tumorigenesis has yet to be investigated. Thus, in the present study, the role of ZNF746 in the invasion and epithelial to mesenchymal transition (EMT) in H460 non-small cell lung cancer (NSCLC) cells was investigated. Invasion assay showed that inhibition of ZNF746 using siRNA transfection inhibited the invasion of H460 NSCLC cells using Boyden chamber. Quantitative PCR (qPCR) analysis revealed that the silencing of ZNF746 attenuated the expression of matrix metalloproteinase (MMP)1, MMP2 and MMP9, but not MMP7, in H460 NSCLC cells. Immunoblotting assay revealed that the expression of E-cadherin and β-catenin of epithelial phenotype was upregulated, while Slug was downregulated in ZNF746 siRNA-transfected H460 NSCLC cells. Accordingly, the mRNA expression of E-cadherin was upregulated while vimentin or Slug, Twist, ZEB as EMT key transcriptional factors were suppressed in ZNF746 siRNA-transfected H460 NSCLC cells. Also, mRNA expression of transcriptional marker Nanog and Octamer-binding transcription factor 4 (OCT4), known to enhance malignancy and metastasis in lung adenocarcinoma, was suppressed in ZNF746 siRNA-transfected H460 NSCLC cells. Notably, the endogenous expression of ZNF746 was induced in parallel with Twist at the protein level during hypoxia. Overall, our findings suggest that inhibition of ZNF746 suppresses the invasion and EMT molecules in H460 NSCLC cells and ZNF746 may be an important target molecule in lung tumorigenesis.