Merck
  • Expression and characterization of a human mitochondrial phenylalanyl-tRNA synthetase.
Expression and characterization of a human mitochondrial phenylalanyl-tRNA synthetase.
Journal of molecular biology (1999-05-18)
J M Bullard, Y C Cai, B Demeler, L L Spremulli
ABSTRACT
Human mitochondrial phenylalanyl-tRNA synthetase (mtPheRS) has been identified from the human EST database. Using consensus sequences derived from conserved regions of the alpha and beta-subunits from bacterial PheRS, two partially sequenced cDNA clones were identified. Unexpectedly, sequence analysis indicated that one of these clones was a truncated form of the other. Detailed analysis indicates that unlike the (alphabeta)2 structure of the prokaryotic and eukaryotic cytoplasmic forms of PheRS, the human mtPheRS consists of a single polypeptide chain. This protein has been cloned and expressed in Escherichia coli. Gel filtration and analytical velocity sedimentation centrifugation indicate that the human mtPheRS is active in a monomeric form. The N-terminal 314 amino acid residues appear to be analogous to the alpha-subunit of the prokaryotic PheRS, while the C-terminal 100 amino acid residues correspond to a region of the beta-subunit known to interact with the anticodon of tRNAPhe. Comparisons with the sequences of PheRS from yeast and Drosophila mitochondria indicate they are 42 % and 51 % identical with the human mtPheRS, respectively. Sequence analysis confirms the presence of motifs characteristic of class II aminoacyl-tRNA synthetases. KM and kcat values for ATP:PPi exchange and for the aminoacylation reaction carried out by human mtPheRS have been determined. Evolutionary origins of this small monomeric human mtPheRS are unknown, however, implications are that this enzyme is a result of the simplification of the more complex (alphabeta)2 bacterial PheRS in which specific functional regions were retained.