Merck
  • Nuclear proteomics with XRCC3 knockdown to reveal the development of doxorubicin-resistant uterine cancer.
Nuclear proteomics with XRCC3 knockdown to reveal the development of doxorubicin-resistant uterine cancer.
Toxicological sciences : an official journal of the Society of Toxicology (2014-03-29)
Jo-Fan Chang, Szu-Ting Lin, Eric Hung, Yi-Ling Lu, Eugenie Wong Soon May, Yi-Wen Lo, Hsiu-Chuan Chou, Hong-Lin Chan
ABSTRACT
The nucleus is a key organelle in mammary cells, which is responsible for several cellular functions including cell proliferation, gene expression, and cell survival. In addition, the nucleus is the primary targets of doxorubicin treatment. In the current study, low-abundance nuclear proteins were enriched for proteomic analysis by using a state-of-the-art two-dimensional differential gel electrophoresis (2D-DIGE) and matrix-assisted laser desorption ionization time-of-flight mass spectrometry (MALDI-TOF MS) strategy to compare and identify the nuclear protein profiling changes responsible for the development of doxorubicin resistance in human uterine cancer cells. The results of the nuclear proteomic analysis indicated that more than 2100 protein features were resolved from an equal pooled amount of three purified nuclear proteins and 117 differentially expressed spots were identified. Of these 117 identified proteins, 48 belonged to nuclear proteins and a positive correlation was observed between the expression levels of 32 of these nuclear proteins and an increase in drug resistance. According to our review of relevant research, nuclear proteins such as DNA repair protein XRCC3 (XRCC3) have not been reported to play roles in the formation of doxorubicin resistance. Previous studies have used RNA interference and cell viability analysis to evidence the essential roles of XRCC3 on its potency in the formation of doxorubicin resistance. To sum up, our nuclear proteomic approaches enabled us to identify numerous proteins, including XRCC3, involved in various drug-resistance-forming mechanisms. Our results provide potential diagnostic markers and therapeutic candidates for treating doxorubicin-resistant uterine cancer.
Sigma-Aldrich
MISSION® esiRNA, targeting human XRCC3
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Xrcc3