Merck
Speckle-type POZ protein, SPOP, is involved in the DNA damage response.
Carcinogenesis (2014-01-24)
Dan Zhang, Haibo Wang, Mianen Sun, Jun Yang, Wei Zhang, Suxia Han, Bo Xu
ABSTRACT
Speckle-type POZ protein (SPOP) is an adaptor of the cullin 3-based ubiquitin ligase responsible for the degradation of oncoproteins frequently overexpressed in many tumor cells. Altered expression and somatic mutations of SPOP have been observed in various tumor types with chromosomal aberrations, indicating a role of SPOP in maintaining genome stability, although a detailed mechanism remains unclear. Here, we show that SPOP is a component of the DNA damage response (DDR). SPOP is recruited to DNA double-strand break sites and it forms nuclear foci after DNA damage. SPOP foci colocalize with γ-H2AX foci and are predominantly dependent on the activity of the ataxia-telangiectasia mutated (ATM) kinase. Furthermore, SPOP interacts with ATM in response to DNA damage. Finally, we demonstrate that knocking down of SPOP resulted in an impaired DDR and a hypersensitivity to ionizing irradiation. Together, we highlight a critical role of SPOP in the DDR.
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Spop
Sigma-Aldrich
MISSION® esiRNA, targeting human SPOP
SAFC
BIS-TRIS
Sigma-Aldrich
BIS-TRIS, Vetec, reagent grade, ≥98%, RNase and DNase free
SAFC
BIS-TRIS
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Pcif1