Merck
  • High-dose diosgenin reduces bone loss in ovariectomized rats via attenuation of the RANKL/OPG ratio.
High-dose diosgenin reduces bone loss in ovariectomized rats via attenuation of the RANKL/OPG ratio.
International journal of molecular sciences (2014-09-27)
Zhiguo Zhang, Changheng Song, Xiaowei Fu, Meijie Liu, Yan Li, Jinghua Pan, Hong Liu, Shaojun Wang, Lihua Xiang, Gary Guishan Xiao, Dahong Ju
ABSTRACT
The aim of this study was to evaluate effect of diosgenin (DG) on rats that had osteoporosis-like features induced by ovariectomy (OVX). Seventy-two six-month-old female Wistar rats were subjected to either ovariectomy (n = 60) or Sham operation (SHAM group, n = 12). Beginning at one week post-ovariectomy, the OVX rats were treated with vehicle (OVX group, n = 12), estradiol valerate (EV group, n = 12), or DG at three doses (DG-L, -M, -H group, n = 12, respectively). After a 12-week treatment, administration of EV or DG-H inhibited OVX-induced weight gain, and administration of EV or DG-H or DG-M had a significantly uterotrophic effect. Bone mineral density (BMD) and indices of bone histomorphometry of tibia were measured. Levels of protein and mRNA expression of osteoprotegerin (OPG) and receptor activator of nuclear factor kappa-B ligand (RANKL) in tibia were evaluated by immunohistochemistry and in situ hybridization. Our results show that DG at a high dose (DG-H) had a significant anti-osteoporotic effect compared to OVX control. DG-H treatment down-regulated expression of RANKL and up-regulated expression of OPG significantly in tibia from OVX rats compared to control, and thus lowered the RANKL/OPG ratio. This suggests that the anti-osteoporotic effect of DG might be associated with modulating the RANKL/OPG ratio and DG had potential to be developed as alternative therapeutic agents of osteoporosis induced by postmenopause.
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
D-(+)-Arabitol, ≥99% (GC)