Merck
  • Preparation of the metabotropic glutamate receptor 5 (mGluR5) PET tracer [(18)F]FPEB for human use: An automated radiosynthesis and a novel one-pot synthesis of its radiolabeling precursor.
Preparation of the metabotropic glutamate receptor 5 (mGluR5) PET tracer [(18)F]FPEB for human use: An automated radiosynthesis and a novel one-pot synthesis of its radiolabeling precursor.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine (2014-10-12)
Keunpoong Lim, David Labaree, Songye Li, Yiyun Huang
ABSTRACT
The radiotracer 3-[(18)F]fluoro-5-(2-pyridinylethynyl)benzonitrile, or [(18)F]FPEB, is a promising PET imaging agent for the metabotropic glutamate subtype 5 receptor (mGluR5). In an effort to develop a routine production method of this radiotracer for use in clinical research we adapted its radiosynthesis to an automated chemistry module. In the meanwhile, we also developed a simplified "one-pot" method for the preparation of the nitrobenzonitrile radiolabeling precursor for [(18)F]FPEB and its reference standard to replace the existing multi-step synthetic approach.
Sigma-Aldrich
2-Ethynylpyridine, 98%