Sigma-Aldrich
Salicylic acid, ≥99%, FG
Pricing and availability is not currently available.
Sigma-Aldrich
Trizma® base, puriss. p.a., ≥99.7% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
Trizma® base, BioUltra, for molecular biology, ≥99.8% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
Chlorotrimethylsilane, ≥98.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Pricing and availability is not currently available.
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
5,7-Dimethoxycoumarin, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
Chlorotrimethylsilane, purified by redistillation, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Pricing and availability is not currently available.
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
Chlorotrimethylsilane, produced by Wacker Chemie AG, Burghausen, Germany, ≥99.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Pricing and availability is not currently available.
Sigma-Aldrich
Chlorotrimethylsilane, puriss., ≥99.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture
Pricing and availability is not currently available.
Supelco
Salicylic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pricing and availability is not currently available.
Supelco
Trizma® base, reference material for titrimetry, certified by BAM, >99.5%
Pricing and availability is not currently available.
Supelco
Melting point standard 235-237°C, analytical standard
Pricing and availability is not currently available.
Supelco
Salicylic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Salicylic acid, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
USP
Salicylic acid, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, anhydrous, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Pricing and availability is not currently available.