Merck
  • Clemizole hydrochloride is a novel and potent inhibitor of transient receptor potential channel TRPC5.
Clemizole hydrochloride is a novel and potent inhibitor of transient receptor potential channel TRPC5.
Molecular pharmacology (2014-08-21)
Julia M Richter, Michael Schaefer, Kerstin Hill
ABSTRACT
Canonical transient receptor potential channel 5 (TRPC5) is a nonselective, Ca(2+)-permeable cation channel that belongs to the large family of transient receptor potential channels. It is predominantly found in the central nervous system with a high expression density in the hippocampus, the amygdala, and the frontal cortex. Several studies confirm that TRPC5 channels are implicated in the regulation of neurite length and growth cone morphology. We identified clemizole as a novel inhibitor of TRPC5 channels. Clemizole efficiently blocks TRPC5 currents and Ca(2+) entry in the low micromolar range (IC50 = 1.0-1.3 µM), as determined by fluorometric intracellular free Ca(2+) concentration ([Ca(2+)]i) measurements and patch-clamp recordings. Clemizole blocks TRPC5 currents irrespectively of the mode of activation, for example, stimulation of G protein-coupled receptors, hypo-osmotic buffer conditions, or by the direct activator riluzole. Electrophysiological whole-cell recordings revealed that the block was mostly reversible. Moreover, clemizole was still effective in blocking TRPC5 single channels in excised inside-out membrane patches, hinting to a direct block of TRPC5 by clemizole. Based on fluorometric [Ca(2+)]i measurements, clemizole exhibits a sixfold selectivity for TRPC5 over TRPC4β (IC50 = 6.4 µM), the closest structural relative of TRPC5, and an almost 10-fold selectivity over TRPC3 (IC50 = 9.1 µM) and TRPC6 (IC50 = 11.3 µM). TRPM3 and M8 as well as TRPV1, V2, V3, and V4 channels were only weakly affected by markedly higher clemizole concentrations. Clemizole was not only effective in blocking heterologously expressed TRPC5 homomers but also TRPC1:TRPC5 heteromers as well as native TRPC5-like currents in the U-87 glioblastoma cell line.
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Clemizole hydrochloride, ≥98% (HPLC)