Merck
  • Jun-regulated genes promote interaction of diffuse large B-cell lymphoma with the microenvironment.
Jun-regulated genes promote interaction of diffuse large B-cell lymphoma with the microenvironment.
Blood (2014-12-24)
Marzenna Blonska, Yifan Zhu, Hubert H Chuang, M James You, Kranthi Kunkalla, Francisco Vega, Xin Lin
ABSTRACT
Diffuse large B-cell lymphoma (DLBCL) is an aggressive disease with a high proliferation rate. However, the molecular and genetic features that drive the aggressive clinical behavior of DLBCL are not fully defined. Here, we have demonstrated that activated Jun signaling is a frequent event in DLBCL that promotes dissemination of malignant cells. Downregulation of Jun dramatically reduces lymphoma cell adhesion to extracellular matrix proteins, subcutaneous tumor size in nude mice, and invasive behavior, including bone marrow infiltration and interaction with bone marrow stromal cells. Furthermore, using a combination of RNA interference and gene expression profiling, we identified Jun target genes that are associated with disseminated lymphoma. Among them, ITGAV, FoxC1, and CX3CR1 are significantly enriched in patients with 2 or more extranodal sites. Our results point to activated Jun signaling as a major driver of the aggressive phenotype of DLBCL.