- Kinetic characterization of sulfasalazine transport by human ATP-binding cassette G2.
Kinetic characterization of sulfasalazine transport by human ATP-binding cassette G2.
Biological & pharmaceutical bulletin (2009-03-03)
Márton Jani, Pál Szabó, Emese Kis, Eva Molnár, Hristos Glavinas, Péter Krajcsi
PMID19252303
ABSTRACT
The pharmacokinetics of sulfasalazine, an anti-inflammatory drug is influenced by ATP-binding cassette G2 (ABCG2) (breast cancer resistance protein (BCRP), mitoxantrone resistance protein (MXR)) both in vitro and clinically. Due to its low passive permeability, the intracellular concentration of sulfasalazine is dependent on uptake transporters, rendering the characterization of transporter specific interactions in cell based experimental systems difficult. Applying membrane assays a detailed kinetic analysis of sulfasalazine ABCG2 interaction was conducted and Km values of 0.70 +/- 0.03 microM and 0.66 +/- 0.08 microM were obtained at pH 7.0 and pH 5.5, respectively.
Sigma-Aldrich
SB-BCRP-M-PREDEASY-ATPase kit, membrane based kit for Human BCRP ATPase Transport Assays