Merck
Roles of CTPL/Sfxn3 and Sfxn family members in pancreatic islet.
Journal of cellular biochemistry (2005-05-03)
Yukako Yoshikumi, Hirosato Mashima, Namiki Ueda, Hideki Ohno, Junko Suzuki, Shigeyasu Tanaka, Michio Hayashi, Nobuo Sekine, Hirohide Ohnishi, Hiroshi Yasuda, Taroh Iiri, Masao Omata, Toshiro Fujita, Itaru Kojima
ABSTRACT
Pancreatic AR42J cells have the feature of pluripotency of the precursor cells of the gut endoderm. Betacellulin (BTC) and activin A (Act) convert them into insulin-secreting cells. Using mRNA differential display techniques, we have identified a novel mitochondrial transporter, which is highly expressed during the course of differentiation, and have designated it citrate transporter protein-like protein (CTPL). Recently sideroflexin 1 (Sfxn1) was shown to be a susceptible gene of flexed-tail (f/f) mice, and CTPL has turned out to be a rat orthologous protein of Sfxn3, a member of sideroflexin family. CTPL/Sfxn3 was targeted to mitochondrial membrane like Sfxn1. The expression levels of CTPL/Sfxn3, Sfxn2, and Sfxn5 were upregulated in the early phase of differentiation into insulin-secreting cells but the expression levels of Sfxn1 and Sfxn3 did not change. All Sfxn family members were expressed in rat pancreatic islet. The expression levels of CTPL/Sfxn3, Sfxn2, and Sfxn5 were also upregulated in islets of streptozotocin-induced diabetic rats compared to normal rats. The downregulation of CTPL/Sfxn3 in a rat insulinoma cell line, INS-1, with the antisense oligonucleotide did not affect the insulin secretion. Taken together, CTPL/Sfxn3 and some other family members might be important in the differentiation of pancreatic beta-cells as a channel or a carrier molecule and be related to the regeneration of pancreatic endocrine cells.