Merck
Activity of recombinant mitogillin and mitogillin immunoconjugates.
The Journal of biological chemistry (1992-08-15)
M Better, S L Bernhard, S P Lei, D M Fishwild, S F Carroll
ABSTRACT
A synthetic gene for the Aspergillus protein toxin mitogillin has been synthesized and expressed in Escherichia coli. The recombinant mitogillin is a potent inhibitor of protein synthesis in vitro with an IC50 of 9.7 pM. Immunoconjugates of recombinant mitogillin derivatized with S-acetylmercaptosuccinic anhydride and 5-methyl-2-iminothiolane modified H65 antibody kill T cell lines and peripheral blood mononuclear cells expressing the human CD5 surface antigen. Native mitogillin contains 4 cysteine residues which form two disulfide pairs (Fernandez-Luna, J. L., Lopez-Otin, C., Soriano, F., and Mendez, E. (1985) Biochemistry 24, 861-867). Three derivatives of mitogillin have been assembled which substitute alanine residues for cysteine residues 5, 147, or 5 and 147. Each of these molecules retains the ability to inhibit protein synthesis in vitro with at most a 2-fold reduction in activity. The derivative mitogillinC147A can be conjugated to 5-methyl-2-iminothiolane- modified H65 antibody directly without pretreatment with S-acetylmercaptosuccinic anhydride, and the immunoconjugate is active against HSB2 cells. Genetic manipulation of toxin genes to expose an accessible cysteine residue into a recombinant product can thus be used to generate immunotoxins without initial derivatization by nonspecific cross-linking reagents.
Sigma-Aldrich
S-Acetylmercaptosuccinic anhydride, 96%