Merck
  • Xylylated dimers of putrescine and polyamines: influence of the polyamine backbone on spermidine transport inhibition.
Xylylated dimers of putrescine and polyamines: influence of the polyamine backbone on spermidine transport inhibition.
Bioorganic & medicinal chemistry letters (2003-09-03)
Laurence Covassin, Michel Desjardins, Denis Soulet, René Charest-Gaudreault, Marie Audette, Richard Poulin
ABSTRACT
Dimeric norspermidine and spermidine derivatives are strong competitive inhibitors of polyamine transport. A xylyl tether was used for the dimerization of various triamines and spermine via a secondary amino group, and of putrescine via an ether or an amino group. Dimerization of putrescine moieties potentiates their ability to compete against spermidine transport to a much greater extent than for triamine dimers.
Sigma-Aldrich
Bis(hexamethylene)triamine, technical grade, 40%