Merck
  • Microwave-assisted preparation of the quorum-sensing molecule 2-heptyl-3-hydroxy-4(1H)-quinolone and structurally related analogs.
Microwave-assisted preparation of the quorum-sensing molecule 2-heptyl-3-hydroxy-4(1H)-quinolone and structurally related analogs.
Nature protocols (2012-05-29)
James T Hodgkinson, Warren R J D Galloway, Martin Welch, David R Spring
ABSTRACT
An optimized procedure for the efficient preparation of 2-heptyl-3-hydroxy-4(1H)-quinolone (Pseudomonas quinolone signal or PQS) and a diverse range of structurally related 2-alkyl-4-quinolones with biological activity is presented. The two-step synthesis begins with the formation of α-chloro ketones by the coupling of a Weinreb amide (2-chloro-N-methoxy-N-methylacetamide) and an appropriate Grignard reagent. The resulting α-chloro ketones can be reacted with commercially available anthranilic acids under microwave irradiation conditions to furnish the desired 2-alkyl-4-quinolone products. As a typical example, the synthesis of PQS, a molecule involved in quorum sensing in the pathogenic bacterium Pseudomonas aeruginosa, is described in detail. The first step of this process (α-chloro ketone formation) takes ∼10 h in total to complete from commercially available bromoheptane and 2-chloro-N-methoxy-N-methylacetamide. The second step (microwave-assisted reaction with anthranilic acid) takes ∼14 h in total to complete (the reaction typically proceeds in ∼30 min, with work-up and purification requiring ∼13 h).
Sigma-Aldrich
2-Chloro-N-methoxy-N-methylacetamide, 98%