Merck
  • Rhodium-catalyzed asymmetric addition of terminal alkynes to diarylphosphinylallenes.
Rhodium-catalyzed asymmetric addition of terminal alkynes to diarylphosphinylallenes.
Chemistry, an Asian journal (2008-05-09)
Takahiro Nishimura, Xun-Xiang Guo, Tamio Hayashi
ABSTRACT
The presence of an acid was found to be essential in the rhodium-catalyzed asymmetric addition of terminal alkynes to diarylphosphinylallenes giving exo-enynes in high yields with high regio- and enantioselectivity. The stereochemical outcome is determined at the protonolysis of the pi-allylrhodium(I) intermediate involved in the catalytic cycle.
Sigma-Aldrich
(Triphenylsilyl)acetylene, 98%