Merck
  • The role of myostatin and the calcineurin-signalling pathway in regulating muscle mass in response to exercise training in the rainbow trout Oncorhynchus mykiss Walbaum.
The role of myostatin and the calcineurin-signalling pathway in regulating muscle mass in response to exercise training in the rainbow trout Oncorhynchus mykiss Walbaum.
The Journal of experimental biology (2005-05-26)
C I Martin, I A Johnston
ABSTRACT
Rainbow trout Oncorhynchus mykiss Walbaum were exercised at 0.8 and 1.6 body lengths s(-1) for 18 h a day over a 30 day period. Exercise resulted in a 24-30% increase in the average cross-sectional area of fast muscle fibres relative to tank-rested controls. The concentrations of growth factors and transcription factors hypothesised to play a role in regulating exercise-induced muscle fibre hypertrophy were measured. Exercise training resulted in a minor increase in calcineurin localisation in the nucleus. However, nuclear factor of T-cells 2 (NFAT2) nuclear localisation did not follow a pattern that was consistent with NFAT2-mediated transcriptional activity and changes in calcineurin signaling. The active peptide of myostatin, a negative regulator of muscle growth in mammals, was downregulated in exercise groups relative to tank-rested controls, but only by 6-7%. It was concluded that myostatin and calcineurin signaling do not play a major role in regulating exercise-induced muscle hypertrophy in trout.