Merck
Integrin alpha 2 I-domain is a binding site for collagens.
Journal of cell science (1995-04-01)
D Tuckwell, D A Calderwood, L J Green, M J Humphries
ABSTRACT
Integrins alpha 1 beta 1 and alpha 2 beta 1 are major cellular receptors for collagens. The alpha 1 and alpha 2 subunits contain a approximately 200 amino acid inserted domain (I-domain) in their N-terminal region and, because of the homology between the I-domains and the collagen-binding A-domains of von Willebrand factor, it has been suggested that the I-domains might mediate the collagen-binding functions of alpha 1 beta 1 and alpha 2 beta 1. In order to fully investigate this hypothesis, we have generated recombinant human alpha 2 I-domain (r alpha 2I) by reverse transcriptase-polymerase chain reaction/bacterial expression and tested its ability to mediate the collagen-binding functions of alpha 2 beta 1. R alpha 2 I binds specifically to type I collagen in a concentration-dependent manner: binding is cation dependent and, like the complete receptor, is supported by magnesium and manganese ions but not by calcium ions. R alpha 2I is recognised by anti-functional anti-alpha 2 monoclonal antibodies 6F1, 5E8 and P1E6 in capture ELISAs, and anti-functional antibodies inhibited r alpha 2I-collagen binding. In addition, r alpha 2I inhibits cell spreading on collagen. R alpha 2I is therefore a collagen-binding domain and can account for many of the collagen-binding functions of integrin alpha 2 beta 1. We have also determined the collagen specificity of r alpha 2I and found that it binds types I, II and XI collagen.