Merck
Aicardi-Goutières syndrome is caused by IFIH1 mutations.
American journal of human genetics (2014-07-06)
Hirotsugu Oda, Kenji Nakagawa, Junya Abe, Tomonari Awaya, Masahide Funabiki, Atsushi Hijikata, Ryuta Nishikomori, Makoto Funatsuka, Yusei Ohshima, Yuji Sugawara, Takahiro Yasumi, Hiroki Kato, Tsuyoshi Shirai, Osamu Ohara, Takashi Fujita, Toshio Heike
ABSTRACT
Aicardi-Goutières syndrome (AGS) is a rare, genetically determined early-onset progressive encephalopathy. To date, mutations in six genes have been identified as etiologic for AGS. Our Japanese nationwide AGS survey identified six AGS-affected individuals without a molecular diagnosis; we performed whole-exome sequencing on three of these individuals. After removal of the common polymorphisms found in SNP databases, we were able to identify IFIH1 heterozygous missense mutations in all three. In vitro functional analysis revealed that IFIH1 mutations increased type I interferon production, and the transcription of interferon-stimulated genes were elevated. IFIH1 encodes MDA5, and mutant MDA5 lacked ligand-specific responsiveness, similarly to the dominant Ifih1 mutation responsible for the SLE mouse model that results in type I interferon overproduction. This study suggests that the IFIH1 mutations are responsible for the AGS phenotype due to an excessive production of type I interferon.
Sigma-Aldrich
Adenosine 5′-Triphosphatase from porcine cerebral cortex, lyophilized powder, ≥0.3 units/mg protein, pH 7.8