Merck
  • Effect of antiretroviral therapy including lopinavir/ritonavir or efavirenz on etonogestrel-releasing implant pharmacokinetics in HIV-positive women.
Effect of antiretroviral therapy including lopinavir/ritonavir or efavirenz on etonogestrel-releasing implant pharmacokinetics in HIV-positive women.
Journal of acquired immune deficiency syndromes (1999) (2014-05-07)
Carolina S Vieira, Maria V Bahamondes, Roberto M de Souza, Milena B Brito, Tatiana R Rocha Prandini, Eliana Amaral, Luis Bahamondes, Geraldo Duarte, Silvana M Quintana, Carolina Scaranari, Rui A Ferriani
ABSTRACT
Data on the interaction between the etonogestrel (ENG) implant and antiretroviral therapy are lacking. We evaluated the effect of 2 highly active antiretroviral therapy (HAART) regimens (1 including efavirenz and the other ritonavir-boosted lopinavir) on the pharmacokinetic (PK) parameters of an ENG-releasing implant in HIV-positive women. Prospective nonrandomized PK study. Forty-five HIV-positive women who desired to use ENG implants were included: 15 had received zidovudine/lamivudine + lopinavir/ritonavir for ≥3 months (LPV/r-based HAART group), 15 had received zidovudine/lamivudine + efavirenz for ≥3 months (EFV-based HAART group), and 15 had not received HAART (non-HAART group). PK parameters were measured using ultra-performance liquid chromatography-mass spectrometry at baseline and 2, 4, 6, 8, 10, 12, 16, 20, and 24 weeks after implant placement. The EFV-based HAART regimen was associated with a reduction in the bioavailability of ENG, which showed decreases of 63.4%, 53.7%, and 70% in the area under the curve (AUC), maximum concentration (Cmax), and minimum concentration (Cmin) of ENG, respectively, compared with the non-HAART group. The LPV/r-based HAART regimen was associated with an increase in ENG bioavailability, which showed 52%, 60.6%, and 33.8% increases in the ENG AUC, Cmax, and Cmin, respectively, compared with the non-HAART group. The coadministration of EFV decreased the bioavailability of ENG released from the implant, which could impair contraceptive efficacy. However, the coadministration of LPV/r increased the bioavailability of ENG released from the implant, which suggests that this antiretroviral combination does not impair the ENG implant efficacy.
Product Description
Desogestrel for system suitability, European Pharmacopoeia (EP) Reference Standard
Desogestrel, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Etonogestrel, ≥98% (HPLC)