Merck
  • MicroRNA-133 modulates the β1-adrenergic receptor transduction cascade.
MicroRNA-133 modulates the β1-adrenergic receptor transduction cascade.
Circulation research (2014-05-09)
Alessandra Castaldi, Tania Zaglia, Vittoria Di Mauro, Pierluigi Carullo, Giacomo Viggiani, Giulia Borile, Barbara Di Stefano, Gabriele Giacomo Schiattarella, Maria Giovanna Gualazzi, Leonardo Elia, Giuliano Giuseppe Stirparo, Maria Luisa Colorito, Gianluigi Pironti, Paolo Kunderfranco, Giovanni Esposito, Marie-Louise Bang, Marco Mongillo, Gianluigi Condorelli, Daniele Catalucci
ABSTRACT
The sympathetic nervous system plays a fundamental role in the regulation of myocardial function. During chronic pressure overload, overactivation of the sympathetic nervous system induces the release of catecholamines, which activate β-adrenergic receptors in cardiomyocytes and lead to increased heart rate and cardiac contractility. However, chronic stimulation of β-adrenergic receptors leads to impaired cardiac function, and β-blockers are widely used as therapeutic agents for the treatment of cardiac disease. MicroRNA-133 (miR-133) is highly expressed in the myocardium and is involved in controlling cardiac function through regulation of messenger RNA translation/stability. To determine whether miR-133 affects β-adrenergic receptor signaling during progression to heart failure. Based on bioinformatic analysis, β1-adrenergic receptor (β1AR) and other components of the β1AR signal transduction cascade, including adenylate cyclase VI and the catalytic subunit of the cAMP-dependent protein kinase A, were predicted as direct targets of miR-133 and subsequently validated by experimental studies. Consistently, cAMP accumulation and activation of downstream targets were repressed by miR-133 overexpression in both neonatal and adult cardiomyocytes following selective β1AR stimulation. Furthermore, gain-of-function and loss-of-function studies of miR-133 revealed its role in counteracting the deleterious apoptotic effects caused by chronic β1AR stimulation. This was confirmed in vivo using a novel cardiac-specific TetON-miR-133 inducible transgenic mouse model. When subjected to transaortic constriction, TetON-miR-133 inducible transgenic mice maintained cardiac performance and showed attenuated apoptosis and reduced fibrosis compared with control mice. miR-133 controls multiple components of the β1AR transduction cascade and is cardioprotective during heart failure.
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate sodium salt monohydrate, ≥98.0% (HPLC), powder
Sigma-Aldrich
(±)-Metoprolol (+)-tartrate salt, ≥98% (titration), powder
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate, ≥98.5% (HPLC), powder
USP
Metoprolol tartrate, United States Pharmacopeia (USP) Reference Standard
Metoprolol tartrate, European Pharmacopoeia (EP) Reference Standard
Supelco
Metoprolol Tartrate, Pharmaceutical Secondary Standard; Certified Reference Material