Merck
  • African histoplasmosis: report of two patients treated with amphotericin B and ketoconazole.
African histoplasmosis: report of two patients treated with amphotericin B and ketoconazole.
Mycoses (1999-01-23)
F C Akpuaka, H C Gugnani, L M Iregbulam
ABSTRACT
Two patients with African histoplasmosis manifested by ulcers and cutaneous swellings are described. The inguinal lymph nodes were also involved in one patient. Treatment with amphotericin B combined with ketoconazole was successful.
Product Description
Ketoconazole, European Pharmacopoeia (EP) Reference Standard
Supelco
Ketoconazole, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Ketoconazole, United States Pharmacopeia (USP) Reference Standard