Merck
[Flecainide acetate].
Annales de cardiologie et d'angeiologie (1996-05-01)
P Bordier, S Garrigue, J Clémenty
ABSTRACT
Flecainide acetate is a powerful antiarrhythmic initially used to treat ventricular arrhythmias. Despite an excess mortality in this indication and in the presence of heart disease, flecainide remains a good antiarrhythmic for supraventricular tachycardias in a healthy heart.
Product Description
Flecainide acetate, European Pharmacopoeia (EP) Reference Standard