Merck
  • Encainide hydrochloride and flecainide acetate: two class 1c antiarrhythmic agents.
Encainide hydrochloride and flecainide acetate: two class 1c antiarrhythmic agents.
Clinical pharmacy (1987-11-01)
S L Chase, G E Sloskey
ABSTRACT
The chemistry, electrophysiology, pharmacokinetics, clinical use and efficacy, adverse effects, drug interactions, and dosage of encainide hydrochloride and flecainide acetate are reviewed. Encainide and flecainide are class 1c antiarrhythmic agents that slow myocardial conduction and mildly prolong the duration of repolarization. Both agents block anterograde conduction over accessory pathways and prolong the effective refractory period of the accessory pathway. Bioavailability of encainide ranges from 7% to 82%, whereas that of flecainide is 90% to 95%. Encainide is metabolized by the liver to two major active metabolites that are slowly eliminated in the urine. About 23% of flecainide's total body clearance is dependent on renal elimination, and drug excretion is slowed in patients with renal dysfunction, requiring dosage adjustments. Both agents are effective in the suppression and prevention of ventricular arrhythmias, including premature ventricular contractions and sustained and nonsustained ventricular tachycardia. These agents may also be valuable in controlling supraventricular arrhythmias. The most common adverse effects of both agents involve the central nervous system and include dizziness, blurred vision, and headache. The potential for proarrhythmic effects is a concern with these agents. The risk is greater in patients with more severe arrhythmias, poor ventricular function, or high serum concentrations of drug. The usual initial oral dosage of encainide hydrochloride is 25 mg three times a day, with a usual dosage range of 100-200 mg/day. Flecainide acetate should be initiated at 100 mg every 12 hours and may be increased up to 400 mg/day. Encainide and flecainide could become useful therapeutic options in the treatment of a variety of arrhythmias.
Product Description
Flecainide acetate, European Pharmacopoeia (EP) Reference Standard