Merck
[5-Fluorocytosine and combinations].
Annales de biologie clinique (1987-01-01)
A Polak
ABSTRACT
The antimycotic and pharmacological properties of 5-fluorocytosine are presented. Especially the aspect of good gastrointestinal absorption, elimination by the kidney, unwanted side effects and appearance of primary and secondary resistance are discussed. The indication of 5 FC monotherapy or combination therapy is shown in various manifestations of deep candidiasis.
Product Description
Flucytosine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
5-Fluorocytosine, Vetec, reagent grade, 99%