Merck
  • Clinical effectiveness of telmisartan alone or in combination therapy for controlling blood pressure and vascular risk in the elderly.
Clinical effectiveness of telmisartan alone or in combination therapy for controlling blood pressure and vascular risk in the elderly.
Clinical interventions in aging (2010-12-15)
Bodh I Jugdutt
ABSTRACT
Elderly patients (age ≥ 65 years) with hypertension are at high risk for vascular complications, especially when diabetes is present. Antihypertensive drugs that inhibit the renin-angiotensin system have been shown to be effective for controlling blood pressure in adult and elderly patients. Importantly, renin-angiotensin system inhibitors were shown to have benefits beyond their classic cardioprotective and vasculoprotective effects, including reducing the risk of new-onset diabetes and associated cardiovascular effects. The discovery that the renin-angiotensin system inhibitor and angiotensin II type 1 (AT(1)) receptor blocker (ARB), telmisartan, can selectively activate the peroxisome proliferator-activated receptor-γ (PPARγ, an established antidiabetic drug target) provides the unique opportunity to prevent and treat cardiovascular complications in high-risk elderly patients with hypertension and new-onset diabetes. Two large clinical trials, ONTARGET (Ongoing Telmisartan Alone in combination with Ramipril Global Endpoint Trial) and TRANSCEND (Telmisartan Randomized AssessmeNt Study in ACE-I iNtolerant subjects with cardiovascular disease) have assessed the cardioprotective and antidiabetic effects of telmisartan. The collective data suggest that telmisartan is a promising drug for controlling hypertension and reducing vascular risk in high-risk elderly patients with new-onset diabetes.
Product Description
Telmisartan, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Telmisartan, ≥98% (HPLC), solid
Telmisartan for peak identification, European Pharmacopoeia (EP) Reference Standard