Merck
Metabolic neutrality in nitrendipine therapy.
Journal of cardiovascular pharmacology (1991-01-01)
M Mancini, T Marotta, L A Ferrara
ABSTRACT
There is evidence that hypertensive patients frequently have other metabolic disorders, such as hyperlipidemia and diabetes mellitus. It is also known that the reduction in high blood pressure alone, disregarding the other cardiovascular risk factors, is unable to reduce mortality to the level of the general population. Moreover, the occurrence of metabolic side effects with some antihypertensive drugs deserves particular attention in the treatment of hypertension. Calcium antagonists seem to be devoid of untoward metabolic effects. In particular, several studies have shown that nitrendipine does not deteriorate glucose tolerance. We have evaluated the effects of nitrendipine on insulin response to i.v. glucose load: no change was observed after 2 months of treatment in both serum insulin levels and glucose percent removal rate in comparison to pretreatment values. No unfavorable change was detectable in the studies aimed at investigating the effects of nitrendipine on lipid metabolism parameters. We observed a 22% increase of the percent removal rate of a lipid emulsion (Intralipid) after nitrendipine (3.11 +/- 1.0 vs. 3.80 +/- 1.0%/min, p less than 0.03). This finding suggests a favorable effect of nitrendipine on triglyceride catabolism, possibly mediated by an interference with lipoprotein lipase activity. The metabolic neutrality of nitrendipine, therefore, leads to considering the usefulness of this drug in an antihypertensive treatment that should not disregard the global risk profile.
Sigma-Aldrich
Nitrendipine, >95%, powder