Merck
Is there any future for felbamate treatment?
Polish journal of pharmacology (2004-06-25)
Kinga K Borowicz, Barbara Piskorska, Zaneta Kimber-Trojnar, Robert Małek, Grzegorz Sobieszek, Stanisław J Czuczwar
ABSTRACT
Felbamate (2-phenyl-1,3-propanediol dicarbamate), a representative of novel antiepileptic drugs (AESs), proved to have broad-spectrum anticonvulsive activity. Particularly beneficial efficacy was found against partial seizures and Lennox-Gastaut syndrome. Therefore, felbamate started to be indicated not only as an adjunctive antiepileptic drug but also in monotherapy. Unfortunately, it was also evidenced that the drug may induce aplastic anemia or hepatic failure. The former complication was frequently described in patients with previously diagnosed hematopoetic disturbances. Thirty-four cases of well-documented bone marrow suppression, occurred fatal in thirteen cases. Subsequently, felbamate's usage was essentially restricted and at present felbamate is not a first-line AED. However, excluding anemia-prone individuals, new possibilities may open for felbamate position in add-on therapy of drug-resistant epilepsy. Experimental studies provide a good theoretical basis for this kind of treatment.