Merck
  • Facilitation of fear extinction by D-cycloserine: theoretical and clinical implications.
Facilitation of fear extinction by D-cycloserine: theoretical and clinical implications.
Learning & memory (Cold Spring Harbor, N.Y.) (2004-10-07)
Rick Richardson, Lana Ledgerwood, Jacquelyn Cranney
ABSTRACT
Anxiety disorders are among the most common psychological disturbances in the industrialized world. Current behavioral therapy procedures for these disorders are somewhat effective, but their efficacy could be substantially improved. Because these procedures are largely based on the process of extinction, manipulations that enhance extinction may lead to improvements in treatment effectiveness. We review the evidence that D-cycloserine (DCS), a partial NMDA agonist, facilitates extinction of learned fear in rats. Although only a few studies have examined the effects of DCS on extinction of learned fear, this work suggests that this drug may have a number of potential clinical benefits. In addition, attempts at interpreting this research illustrate our limited understanding of the processes involved in extinction.
Sigma-Aldrich
D-Cycloserine
Sigma-Aldrich
D-Cycloserine, synthetic