Merck
  • The effects of cyproterone acetate on sleeping and waking penile erections in pedophiles: possible implications for treatment.
The effects of cyproterone acetate on sleeping and waking penile erections in pedophiles: possible implications for treatment.
Canadian journal of psychiatry. Revue canadienne de psychiatrie (1992-02-01)
A J Cooper, Z Cernovovsky
ABSTRACT
This study reports the short term effects in five pedophiles of the antiandrogenic drug cyproterone acetate (CPA) on nocturnal penile tumescence (NPT); penile responses to erotic stimuli in the laboratory; and sex hormones (testosterone, LH, FSH and prolactin). During the administration of CPA, NPT, laboratory arousal and hormone measures (except prolactin) all decreased. Waking laboratory measures were influenced less and were more variable (one subject showed greater arousal) than NPT measures. The changes in NPT closely paralleled the reduction in testosterone. The results are discussed with reference to the known psycho-neuroendocrinology of sleeping and waking erections.
Sigma-Aldrich
Cyproterone acetate, ≥98%
Cyproterone acetate, European Pharmacopoeia (EP) Reference Standard