Merck
  • Total syntheses of leuconoxine, leuconodine B, and melodinine E by oxidative cyclic aminal formation and diastereoselective ring-closing metathesis.
Total syntheses of leuconoxine, leuconodine B, and melodinine E by oxidative cyclic aminal formation and diastereoselective ring-closing metathesis.
Organic letters (2014-04-24)
Atsushi Umehara, Hirofumi Ueda, Hidetoshi Tokuyama
ABSTRACT
Total syntheses of leuconodine B, melodinine E, and leuconoxine were accomplished via a divergent route. The [5.5.6.6]diazafenestrane skeleton was constructed from an indole-3-acetamide derivative via DMDO oxidation to hydroxylindolenine, TMSOTf/2,6-lutidine mediated cyclic aminal formation, and diastereoseletive ring-closing metathesis of a triene derivative.
Sigma-Aldrich
Indole-3-acetamide, 98%