Merck
  • Evidence for the HOOO(-) anion in the ozonation of 1,3-dioxolanes: hemiortho esters as the primary products.
Evidence for the HOOO(-) anion in the ozonation of 1,3-dioxolanes: hemiortho esters as the primary products.
Journal of the American Chemical Society (2002-09-19)
Bozo Plesnicar, Janez Cerkovnik, Tell Tuttle, Elfi Kraka, Dieter Cremer
ABSTRACT
Low-temperature ozonation (-78 degrees C) of 2-methyl-1,3-dioxolane (1a) in acetone-d6, methyl acetate, and tert-butyl methyl ether produced both the corresponding acetal hydrotrioxide (3a, ROOOH) and the hemiortho ester (2a, ROH) in molar ratio 1:5. Both intermediates were fully characterized by 1H, 13C, and 17O NMR spectroscopy, and they both decomposed to the corresponding hydroxy ester at higher temperatures. The mechanism involving the HOOO- anion formed by the abstraction of the hydride ion by ozone to form an ion pair, R+ -OOOH, with its subsequent collapse to either the corresponding hemiortho ester (ROH) or the acetal hydrotrioxide (ROOOH) was proposed. This mechanism is supported by the PISA/B3LYP/6-311++G(3df,3pd) calculations.
Sigma-Aldrich
2-Methyl-1,3-dioxolane, 97%