Merck
  • Characterization of plasma lipoproteins separated and purified by agarose-column chromatography.
Characterization of plasma lipoproteins separated and purified by agarose-column chromatography.
The Biochemical journal (1974-04-01)
L L Rudel, J A Lee, M D Morris, J M Felts
ABSTRACT
1. A simple method for isolation of individual human plasma lipoprotein classes is presented. In this technique, lipoproteins are removed from plasma at d1.225 by ultracentrifugation, after which they are separated and purified by agarose-column chromatography. 2. Three major classes are obtained after agarose-column chromatography. Separation between classes is excellent; more than 95% of the lipoproteins eluted from the column are recovered in the form of a purified lipoprotein class. 3. Each lipoprotein class was characterized immunologically, chemically, electrophoretically and by electron microscopy. A comparison of the properties of the column-isolated lipoproteins was made with very-low-density lipoproteins, low-density lipoproteins, and high-density lipoproteins separated by sequential ultracentrifugation at densities of 1.006, 1.063 and 1.21 respectively. 4. By each criterion, peak-I lipoproteins from the agarose column are the same as very-low-density lipoproteins, peak-II lipoproteins are the same as low-density lipoproteins, and peak-III lipoproteins are the same as high-density lipoproteins. Thus the lipoprotein classes isolated by both methods are similar if not identical. 5. The agarose-column separation technique offers the advantage of a two- to three-fold saving in time. In addition, the column-elution pattern serves as a recording of the size distribution of lipoproteins in plasma. 6. The most complete characterization is reported for human plasma lipoproteins. The results with rhesus-monkey and rabbit lipoproteins were identical.
Sigma-Aldrich
Lipoprotein, low density from human plasma, ≥95% (SDS-PAGE), solution