Merck
  • Faster decomposition under increased atmospheric CO₂ limits soil carbon storage.
Faster decomposition under increased atmospheric CO₂ limits soil carbon storage.
Science (New York, N.Y.) (2014-04-26)
Kees Jan van Groenigen, Xuan Qi, Craig W Osenberg, Yiqi Luo, Bruce A Hungate
ABSTRACT
Soils contain the largest pool of terrestrial organic carbon (C) and are a major source of atmospheric carbon dioxide (CO2). Thus, they may play a key role in modulating climate change. Rising atmospheric CO2 is expected to stimulate plant growth and soil C input but may also alter microbial decomposition. The combined effect of these responses on long-term C storage is unclear. Combining meta-analysis with data assimilation, we show that atmospheric CO2 enrichment stimulates both the input (+19.8%) and the turnover of C in soil (+16.5%). The increase in soil C turnover with rising CO2 leads to lower equilibrium soil C stocks than expected from the rise in soil C input alone, indicating that it is a general mechanism limiting C accumulation in soil.
Sigma-Aldrich
Carbon dioxide, ≥99.8%
Sigma-Aldrich
Carbon-12C dioxide, 99.99 atom % 12C
Sigma-Aldrich
Carbon-12C dioxide, 99.9 atom % 12C