- Structure-activity relationship studies on new DABOS: effect of substitutions at pyrimidine C-5 and C-6 positions on anti-HIV-1 activity.
Structure-activity relationship studies on new DABOS: effect of substitutions at pyrimidine C-5 and C-6 positions on anti-HIV-1 activity.
Antiviral chemistry & chemotherapy (2001-07-05)
G Sbardella, A Mai, M Artico, P Chimenti, S Massa, R Loddo, M E Marongiu, P La Collat, A Pani
PMID11437321
ABSTRACT
Several 5-alkyl, 5-alkenyl, 5-iso-alkyl, 5-halo, 5-aminomethyl and 5-carboxy derivatives of S-DABOs (dihydro-alkyl (or cyclo-alkyl)thio-benzyloxopyrimidines), DATNOs (dihydro-alkylthionaphthylmethyl-oxopyrimidines) and F2-S-DABOs (dihydro-alkyl (or cyclo-alkyl)thio-2,6-difluorobenzyl-oxopyrimidines) have been prepared and tested as anti-HIV-1 agents. S-DABO derivatives bearing at C-6 position monosubstituted phenylmethyl or heteroarylmethyl units have also been synthesized. 2-Alkylthio-3,4-dihydropyrimidin-4(3H)-one derivatives of F2-S-DABO series bearing small alkyl groups at C-5 proved to be potent inhibitors of HIV-1 replication in vitro with selectivity indexes ranging from 250 to >2,500.