Merck
  • Synthesis of novel inhibitors of the HIV-1 protease: difunctional enols of simple N-protected amino acids.
Synthesis of novel inhibitors of the HIV-1 protease: difunctional enols of simple N-protected amino acids.
Bioorganic & medicinal chemistry (1994-05-01)
M Vaillancourt, B Vanasse, N Le Berre, E Cohen, G Sauvé
ABSTRACT
A series of enol HIV-1 protease inhibitors which show competitive inhibition and the structure-activity relationship study which led to the design of these compounds are reported. By systematically modifying simple amino acids, Boc-Phe enol and Boc-Tyr enol derivatives yield nanomolar Kiapp values (Kiapp = 0.485 microM and Kiapp = 0.425 microM, respectively). These enols are of low molecular weight (< 500 g/mol) and of non-peptidic nature. The enols are synthesized in a one step chemical synthesis and modifications to increase their potency could easily be performed. Boc-Phe enol and Boc-Tyr enol showed low inhibitory effect on pepsin, Kiapps of 23 and 149 microM, respectively, and Boc-Phe enol showed a Kiapp of 20 microM for cathepsin D. Neither of these two compounds inhibited renin (< 10% inhibition at 200 microM).