Merck
Fragrance material review on cyclohexyl acetate.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2008-10-11)
S P Bhatia, C S Letizia, A M Api
ABSTRACT
A toxicologic and dermatologic review of cyclohexyl acetate when used as a fragrance ingredient is presented.
Sigma-Aldrich
Cyclohexyl acetate, 99%