Merck
  • Bioavailability of bacampicillin and talampicillin, two oral prodrugs of ampicillin.
Bioavailability of bacampicillin and talampicillin, two oral prodrugs of ampicillin.
Antimicrobial agents and chemotherapy (1981-12-01)
J Sjövall, L Magni, E Vinnars
ABSTRACT
A 200-mg amount of bacampicillin showed a significantly higher relative extent of bioavailability than did a 250-mg amount of talampicillin, possibly due to their different stability in the digestive juices.