Merck
  • Efficacy and safety of vildagliptin and voglibose in Japanese patients with type 2 diabetes: a 12-week, randomized, double-blind, active-controlled study.
Efficacy and safety of vildagliptin and voglibose in Japanese patients with type 2 diabetes: a 12-week, randomized, double-blind, active-controlled study.
Diabetes, obesity & metabolism (2010-07-02)
Y Iwamoto, A Kashiwagi, N Yamada, S Terao, N Mimori, M Suzuki, H Tachibana
ABSTRACT
To confirm the efficacy of vildagliptin in patients with type 2 diabetes (T2D) by testing the hypothesis that glycosylated haemoglobin (HbA1c) reduction with vildagliptin is superior to that with voglibose after 12 weeks of treatment. In this 12-week, randomized, double-blind, active-controlled, parallel-group study, the efficacy and safety of vildagliptin (50 mg bid, n = 188) was compared with that of voglibose (0.2 mg tid, n = 192) in patients with T2D who were inadequately controlled with diet and exercise. The characteristics of two groups were well matched at baseline. The mean age, body mass index (BMI) and HbA1c were 59.1 years, 24.9 kg/m(2) and 7.6%, respectively. At baseline, fasting plasma glucose (FPG) and 2-h postprandial glucose (PPG) were 9.01 mmol/l (162.2 mg/dl) and 13.57 mmol/l (244.3 mg/dl), respectively. The adjusted mean change in HbA1c from baseline to endpoint was -0.95 +/- 0.04% in the vildagliptin-treated patients and -0.38 +/- 0.04% in those receiving voglibose (between-group change = 0.57 +/- 0.06%, 95% confidence interval (CI) (-0.68 to -0.46%), p < 0.001), showing that vildagliptin was superior to voglibose. Endpoint HbA1c < or = 6.5% was achieved in 51% vildagliptin-treated patients compared with 24% patients who were on voglibose (p < 0.001). Vildagliptin also exhibited significantly (p < 0.001) greater reduction compared with voglibose in both FPG [1.34 vs. 0.43 mmol/l (24.1 vs. 7.8 mg/dl)] and 2-h PPG [2.86 vs. 1.1 mmol/l (51.5 vs. 19.8 mg/dl)]. Overall adverse events (AEs) were lower in the vildagliptin-treated patients compared with that in the voglibose-treated patients (61.2 vs. 71.4%), with no incidence of hypoglycaemia and serious adverse events with vildagliptin. Gastrointestinal AEs were significantly lower with vildagliptin compared with that of the voglibose (18.6 vs. 32.8%; p = 0.002). Vildagliptin (50 mg bid) showed superior efficacy and better tolerability compared with voglibose in Japanese patients with T2D.
Sigma-Aldrich
Voglibose, ≥97.0% (TLC)